Ferric oleate ≥95%

Supplier: Ambeed
Warning

A990714-5G A990714-100G A990714-25G
77599-412EA 41 USD
77599-412 77623-390 77784-666
Ferric oleate ≥95%
Ferric oleate

Precaución: for Research Use Only

Formula: C₅₄H₉₉FeO₆
MW: 900.21 g/mol
MDL Number: MFCD00067235
CAS Number: 1120-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR