(3-Oxo-6-oxido-2,4,5,7-tetraiodo-3h-xanthen-9-yl)-3,4,5,6-tetrachlorobenzoate sodium salt ≥95%

Supplier: Ambeed
Warning

A990690-1G A990690-5G A990690-100G A990690-25G
77599-408EA 6.15 USD
77599-408 77599-410 77623-388 77784-665
(3-Oxo-6-oxido-2,4,5,7-tetraiodo-3h-xanthen-9-yl)-3,4,5,6-tetrachlorobenzoate sodium salt ≥95%
(3-Oxo-6-oxido-2,4,5,7-tetraiodo-3h-xanthen-9-yl)-3,4,5,6-tetrachlorobenzoate sodium salt

Precaución: for Research Use Only

MDL Number: MFCD00005043
CAS Number: 11121-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR