(S)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Desmethyl Levofloxacin
A990563-2MG
A990563-5MG
A990563-10MG
77623-376EA
42.23
USD
77623-376
77623-378
77623-374
(S)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid ≥98%
(S)-9-Fluoro-3-methyl-7-oxo-10-(piperazin-1-yl)-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₇H₁₈FN₃O₄ MW: 347.34 g/mol |
MDL Number:
MFCD15071265 CAS Number: 117707-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...