Sodium dihydrogen citrate ≥99%

Supplier: Ambeed
Warning

A990484-1000G A990484-100G A990484-25G A990484-500G A990484-5G
77595-590EA 29.73 USD
77595-590 77595-592 77595-594 77595-596 77873-405
Sodium dihydrogen citrate ≥99%
Sodium dihydrogen citrate

Precaución: for Research Use Only

Formula: C₆H₇NaO₇
MDL Number: MFCD00013067
CAS Number: 18996-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR