Methyl 5-amino-2-bromo-3-fluorobenzoate ≥95%

Supplier: Ambeed
Warning

A990100-1G A990100-100MG A990100-250MG
77573-968EA 163 USD
77573-968 77784-655 77784-656
Methyl 5-amino-2-bromo-3-fluorobenzoate ≥95%
Methyl 5-amino-2-bromo-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrFNO₂
MDL Number: MFCD24119832
CAS Number: 1036389-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR