6-(4-([1,1'-Biphenyl]-4-yl)-5-((benzyloxy)carbonyl)-6-methyl-2-oxo-3,4-dihydropyrimidin-1(2H)-yl)hexanoic acid ≥95%
Supplier: Ambeed
|
Danger
|
Synonyms:
116-9e
A989751-10MG
A989751-1MG
A989751-5MG
77599-396EA
96.35
USD
77599-396
77599-398
77599-400
6-(4-([1,1'-Biphenyl]-4-yl)-5-((benzyloxy)carbonyl)-6-methyl-2-oxo-3,4-dihydropyrimidin-1(2H)-yl)hexanoic acid ≥95%
6-(4-([1,1'-Biphenyl]-4-yl)-5-((benzyloxy)carbonyl)-6-methyl-2-oxo-3,4-dihydropyrimidin-1(2H)-yl)hexanoic acid
Precaución: for Research Use Only
Formula:
C₃₁H₃₂N₂O₅ MW: 512.6 g/mol |
MDL Number:
MFCD18632539 CAS Number: 831217-43-7 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...