4-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid 95+%

Supplier: Ambeed
Warning

A989590-1G A989590-250MG
77884-911EA 260 USD
77884-911 77884-912
4-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid 95+%
4-(Methoxycarbonyl)-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BF₃O₄
MDL Number: MFCD13181653
CAS Number: 1451375-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR