5-Ethynyl-2-methoxypyrimidine ≥95%

Supplier: Ambeed
Warning

A989269-1G A989269-250MG A989269-100MG
77784-643EA 582 USD
77784-643 77784-644 77784-642
5-Ethynyl-2-methoxypyrimidine ≥95%
5-Ethynyl-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MDL Number: MFCD13189784
CAS Number: 1059705-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR