tert-Butyl 2-(tert-butyldimethylsilyloxy) ethylcarbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (2-((tert-butyldimethylsilyl)oxy)ethyl)carbamate

A989168-5G A989168-1G A989168-100MG A989168-250MG
77573-966EA 742 USD
77573-966 77623-356 77784-639 77784-640
tert-Butyl 2-(tert-butyldimethylsilyloxy) ethylcarbamate ≥95%
tert-Butyl 2-(tert-butyldimethylsilyloxy) ethylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₉NO₃Si
CAS Number: 203738-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR