Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)-2-hydroxypropane-1-sulfonate ≥99%

Supplier: Ambeed
Danger

Synonyms: TAPSO sodium

A989037-25G A989037-5G
77618-710EA 22.66 USD
77618-710 77618-712
Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)-2-hydroxypropane-1-sulfonate ≥99%
Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)-2-hydroxypropane-1-sulfonate

Precaución: for Research Use Only

Formula: C₇H₁₆NNaO₇S
MW: 281.25 g/mol
MDL Number: MFCD00070009
CAS Number: 105140-25-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR