Ethyl ethanesulfonate ≥95%

Supplier: Ambeed
Warning

A989025-1G A989025-25G A989025-250MG A989025-5G
77599-390EA 21.53 USD
77599-390 77784-631 77784-630 77784-632
Ethyl ethanesulfonate ≥95%
Ethyl ethanesulfonate

Precaución: for Research Use Only

Formula: C₄H₁₀O₃S
MDL Number: MFCD01728649
CAS Number: 1912-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR