2-(tert-Butoxy)butane ≥95%

Supplier: Ambeed
Danger

A989018-1G A989018-250MG
77623-352EA 93.73 USD
77623-352 77623-354
2-(tert-Butoxy)butane ≥95%
2-(tert-Butoxy)butane

Precaución: for Research Use Only

Formula: C₈H₁₈O
MDL Number: MFCD23160711
CAS Number: 32970-45-9
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR