17-Bromoheptadecanoic acid ≥98%

Supplier: Ambeed
Warning

A988995-1G A988995-250MG A988995-5G
77599-384EA 89.18 USD
77599-384 77599-386 77599-388
17-Bromoheptadecanoic acid ≥98%
17-Bromoheptadecanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₃₃BrO₂
MW: 349.347 g/mol
MDL Number: MFCD00040782
CAS Number: 13099-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR