Benzyl (5-bromopentyl)carbamate ≥95%

Supplier: Ambeed
Danger

A988924-1G A988924-250MG A988924-5G A988924-100MG
77599-382EA 97.38 USD
77599-382 77623-346 77623-348 77784-626
Benzyl (5-bromopentyl)carbamate ≥95%
Benzyl (5-bromopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
MW: 300.1915 g/mol
MDL Number: MFCD12974655
CAS Number: 161533-09-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR