2-Bromo-4-(difluoromethyl)-5-fluoropyridine ≥95%

Supplier: Ambeed
Warning

A988881-250MG A988881-100MG
77784-625EA 169 USD
77784-625 77784-624
2-Bromo-4-(difluoromethyl)-5-fluoropyridine ≥95%
2-Bromo-4-(difluoromethyl)-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MDL Number: MFCD25477691
CAS Number: 1805299-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR