3-Bromo-4-methylbenzamide 95+%

Supplier: Ambeed
Warning

A988840-25G A988840-5G
77882-860EA 284 USD
77882-860 77882-861
3-Bromo-4-methylbenzamide 95+%
3-Bromo-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD03306087
CAS Number: 183723-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR