1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborole-7-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A988753-100MG A988753-1G
77623-338EA 264.71 USD
77623-338 77623-340
1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborole-7-carboxylic acid ≥95%
1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇BO₄
MW: 177.95 g/mol
MDL Number: MFCD20661020
CAS Number: 1268335-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR