5-(Chloromethyl)-1-methyl-2-nitro-1H-imidazole ≥95%

Supplier: Ambeed
Danger

A988648-250MG A988648-100MG
77784-620EA 147 USD
77784-620 77784-619
5-(Chloromethyl)-1-methyl-2-nitro-1H-imidazole ≥95%
5-(Chloromethyl)-1-methyl-2-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₅H₆ClN₃O₂
MDL Number: MFCD09907691
CAS Number: 87544-76-1
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR