1-(Benzo[d]thiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A988378-10MG A988378-5MG A988378-100MG A988378-250MG A988378-50MG
77684-917EA 94 USD
77684-917 77684-918 77893-779 77893-780 77893-781
1-(Benzo[d]thiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol ≥95%, Ambeed.Inc
1-(Benzo[d]thiazol-2-yl)-1-(2,4-dimethylphenyl)ethanol

Precaución: For research use only.

Formula: C₁₇H₁₇NOS
MDL Number: MFCD20527324
CAS Number: 1253901-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR