4-(4-Hydroxy-3-methoxyphenethyl)-2,6-dimethoxyphenol ≥98%

Supplier: Ambeed
Warning

Synonyms: Dendrophenol

A987385-10MG A987385-1MG A987385-5MG
77599-374EA 272.65 USD
77599-374 77599-376 77599-378
4-(4-Hydroxy-3-methoxyphenethyl)-2,6-dimethoxyphenol ≥98%
4-(4-Hydroxy-3-methoxyphenethyl)-2,6-dimethoxyphenol

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₅
MW: 304.34 g/mol
MDL Number: MFCD27989278
CAS Number: 108853-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR