Biotin-PEG2-Acid ≥95%

Supplier: Ambeed
Warning

A987353-1G A987353-50MG A987353-100MG A987353-250MG
77784-588EA 332 USD
77784-588 77784-590 77784-587 77784-589
Biotin-PEG2-Acid ≥95%
Biotin-PEG2-Acid

Precaución: for Research Use Only

Formula: C₁₇H₂₉N₃O₆S
MDL Number: MFCD22056305
CAS Number: 1365655-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR