5-Amino-N-(3-fluoro-4-methoxybenzyl)-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide ≥95%

Supplier: Ambeed
Warning

Synonyms: VU0467485

A987343-10MG A987343-5MG
77599-370EA 102.5 USD
77599-370 77599-372
5-Amino-N-(3-fluoro-4-methoxybenzyl)-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide ≥95%
5-Amino-N-(3-fluoro-4-methoxybenzyl)-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₁₇FN₄O₂S
MDL Number: MFCD31415018
CAS Number: 1451994-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR