1,3-Diphenylguanidine ≥98%

Supplier: Ambeed
Danger

A987302-100G A987302-25G A987302-500G
77618-702EA 6.18 USD
77618-702 77618-704 77747-737
1,3-Diphenylguanidine ≥98%
1,3-Diphenylguanidine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₃
MDL Number: MFCD00001758
CAS Number: 102-06-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR