3-Hexylundecan-1-ol ≥95%

Supplier: Ambeed
Warning

A987177-250MG A987177-100MG
77784-586EA 120 USD
77784-586 77784-585
3-Hexylundecan-1-ol ≥95%
3-Hexylundecan-1-ol

Precaución: for Research Use Only

Formula: C₁₇H₃₆O
MDL Number: MFCD28359609
CAS Number: 51655-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR