Fluoromethyl 2-pyridyl sulfone ≥95%

Supplier: Ambeed
Danger

A986778-1G A986778-250MG
77784-576EA 76 USD
77784-576 77784-577
Fluoromethyl 2-pyridyl sulfone ≥95%
Fluoromethyl 2-pyridyl sulfone

Precaución: for Research Use Only

Formula: C₆H₆FNO₂S
MDL Number: MFCD28100935
CAS Number: 1365765-53-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR