Ethyl 7-oxoheptanoate ≥95%

Supplier: Ambeed
Warning

A986700-1G A986700-250MG A986700-100MG
77623-318EA 133.9 USD
77623-318 77623-320 77784-574
Ethyl 7-oxoheptanoate ≥95%
Ethyl 7-oxoheptanoate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
MDL Number: MFCD07186258
CAS Number: 3990-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR