Amino-PEG4-CH2CO2tBu ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 14-amino-3,6,9,12-tetraoxatetradecanoate, Amino-PEG4-C1-Boc

A986402-1G A986402-250MG A986402-100MG A986402-5G
77573-952EA 119.48 USD
77573-952 77599-368 77623-310 77623-312
Amino-PEG4-CH2CO2tBu ≥95%
Amino-PEG4-CH2CO2tBu

Precaución: for Research Use Only

Formula: C₁₄H₂₉NO₆
MW: 307.383 g/mol
MDL Number: MFCD28122943
CAS Number: 864680-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR