Cyclopropylacetyl chloride ≥98%

Supplier: Ambeed
Danger

A986355-5G A986355-250MG A986355-1G
77573-948EA 483 USD
77573-948 77784-566 77784-565
Cyclopropylacetyl chloride ≥98%
Cyclopropylacetyl chloride

Precaución: for Research Use Only

Formula: C₅H₇ClO
MDL Number: MFCD11110393
CAS Number: 54322-65-5
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR