Proparyl-PEG4-methane ≥96%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG3-methane, 2,5,8,11-Tetraoxatetradec-13-yne

A986257-1G A986257-250MG A986257-5G
77623-304EA 100.94 USD
77623-304 77784-555 77784-556
Proparyl-PEG4-methane ≥96%
Proparyl-PEG4-methane

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MDL Number: MFCD27977518
CAS Number: 89635-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR