tert-Butyl 1-(tosyloxy)-3,6,9,12,15-pentaoxaoctadecan-18-oate ≥95%

Supplier: Ambeed
Warning

Synonyms: Tos-PEG5-C2-Boc

A986251-100MG A986251-1G A986251-250MG A986251-5G
77623-296EA 9.27 USD
77623-296 77623-298 77623-300 77623-302
tert-Butyl 1-(tosyloxy)-3,6,9,12,15-pentaoxaoctadecan-18-oate ≥95%
tert-Butyl 1-(tosyloxy)-3,6,9,12,15-pentaoxaoctadecan-18-oate

Precaución: for Research Use Only

Formula: C₂₄H₄₀O₁₀S
MW: 520.6334 g/mol
MDL Number: MFCD22683305
CAS Number: 850090-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR