N-(2-((2-Methoxyphenyl)amino)-4'-methyl-[4,5'-bithiazol]-2'-yl)acetamide ≥98%

Supplier: Ambeed
Warning

A984976-250MG A984976-100MG A984976-50MG
77623-278EA 393.46 USD
77623-278 77784-543 77784-544
N-(2-((2-Methoxyphenyl)amino)-4'-methyl-[4,5'-bithiazol]-2'-yl)acetamide ≥98%
N-(2-((2-Methoxyphenyl)amino)-4'-methyl-[4,5'-bithiazol]-2'-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₄O₂S₂
MDL Number: MFCD02045222
CAS Number: 315705-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR