(S)-2-(6-(4-Chlorophenyl)-1-methyl-8-(1-methyl-1H-pyrazol-4-yl)-4H-benzo[c]isoxazolo[4,5-e]azepin-4-yl)acetamide ≥98%

Supplier: Ambeed
Warning

Synonyms: BET bromodomain inhibitor

A984974-10MG A984974-1MG A984974-5MG
77599-358EA 115.83 USD
77599-358 77599-360 77599-362
(S)-2-(6-(4-Chlorophenyl)-1-methyl-8-(1-methyl-1H-pyrazol-4-yl)-4H-benzo[c]isoxazolo[4,5-e]azepin-4-yl)acetamide ≥98%
(S)-2-(6-(4-Chlorophenyl)-1-methyl-8-(1-methyl-1H-pyrazol-4-yl)-4H-benzo[c]isoxazolo[4,5-e]azepin-4-yl)acetamide

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClN₅O₂
MW: 445.9 g/mol
MDL Number: MFCD28144506
CAS Number: 1505453-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR