N-(5-Fluoro-4-methoxy-2-nitrophenyl)acetamide ≥98%

Supplier: Ambeed
Danger

A984803-5G A984803-1G
77784-542EA 115 USD
77784-542 77784-541
N-(5-Fluoro-4-methoxy-2-nitrophenyl)acetamide ≥98%
N-(5-Fluoro-4-methoxy-2-nitrophenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₉FN₂O₄
MDL Number: MFCD22053302
CAS Number: 448-26-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR