4-Bromo-N-(5-fluoro-2-methylphenyl)benzenesulfonamide ≥95%

Supplier: Ambeed
Warning

A984495-1G A984495-5G
77784-524EA 20 USD
77784-524 77784-525
4-Bromo-N-(5-fluoro-2-methylphenyl)benzenesulfonamide ≥95%
4-Bromo-N-(5-fluoro-2-methylphenyl)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrFNO₂S
MDL Number: MFCD05139658
CAS Number: 885396-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR