2-(2-(2-(Benzyloxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate ≥95%

Supplier: Ambeed
Warning

A984379-1G A984379-250MG A984379-5G
77595-584EA 83.03 USD
77595-584 77603-392 77599-348
2-(2-(2-(Benzyloxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate ≥95%
2-(2-(2-(Benzyloxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₀H₂₆O₆S
MW: 394.4819 g/mol
MDL Number: MFCD31555345
CAS Number: 84131-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR