4-(2-Chloroethyl)-1H-pyrazole ≥98%, Ambeed.Inc

Supplier: Ambeed
Danger

A984303-250MG A984303-100MG A984303-1G
77684-914EA 20 USD
77684-914 77891-736 77891-737
4-(2-Chloroethyl)-1H-pyrazole ≥98%, Ambeed.Inc
4-(2-Chloroethyl)-1H-pyrazole

Precaución: For research use only.

Formula: C₅H₇ClN₂
MDL Number: MFCD08059907
CAS Number: 438475-37-7
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR