PF-04965842 ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(cis-3-(Methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclobutyl)propane-1-sulphonamide, Abrocitinib

A983986-1MG A983986-5MG A983986-10MG
77599-344EA 54.33 USD
77599-344 77599-346 77599-342
PF-04965842 ≥98%
PF-04965842

Precaución: for Research Use Only

Formula: C₁₄H₂₁N₅O₂S
MW: 323.42 g/mol
MDL Number: MFCD30187577
CAS Number: 1622902-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR