9,9-Dimethyl-9H-fluorene-2,7-diamine ≥97%

Supplier: Ambeed
Warning

A983761-5G A983761-250MG
77784-516EA 57 USD
77784-516 77784-515
9,9-Dimethyl-9H-fluorene-2,7-diamine ≥97%
9,9-Dimethyl-9H-fluorene-2,7-diamine

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂
MDL Number: MFCD30293216
CAS Number: 216454-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR