1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12-tetraoxapentadecan-15-oic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Pomalidomide-PEG4-COOH

A983074-50MG A983074-100MG A983074-1G
77595-574EA 117.88 USD
77595-574 77599-338 77623-262
1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12-tetraoxapentadecan-15-oic acid ≥95%
1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12-tetraoxapentadecan-15-oic acid

Precaución: for Research Use Only

Formula: C₂₄H₃₁N₃O₁₀
MW: 521.52 g/mol
MDL Number: MFCD32173789
CAS Number: 2138440-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR