(1R,3R,4R,7S)-3-(4-Benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite ≥97%

Supplier: Ambeed
Warning

Synonyms: DMT-locMeC(bz) Phosphoramidite

A983073-1G A983073-100MG A983073-250MG A983073-5G
77623-260EA 250.29 USD
77623-260 77784-503 77784-504 77784-505
(1R,3R,4R,7S)-3-(4-Benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite ≥97%
(1R,3R,4R,7S)-3-(4-Benzamido-5-methyl-2-oxopyrimidin-1(2H)-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite

Precaución: for Research Use Only

Formula: C₄₈H₅₄N₅O₉P
MW: 875.94 g/mol
CAS Number: 206055-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR