(1R,3R,4R,7S)-3-(6-Benzamido-9H-purin-9-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite ≥97%
Supplier: Ambeed
|
Warning
|
A983045-100MG
A983045-1G
A983045-250MG
A983045-5G
77623-254EA
59.74
USD
77623-254
77623-256
77623-258
77784-501
(1R,3R,4R,7S)-3-(6-Benzamido-9H-purin-9-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite ≥97%
(1R,3R,4R,7S)-3-(6-Benzamido-9H-purin-9-yl)-1-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-2,5-dioxabicyclo[2.2.1]heptan-7-yl (2-cyanoethyl) diisopropylphosphoramidite
Precaución: for Research Use Only
Formula:
C₄₈H₅₂N₇O₈P |
MDL Number:
MFCD31707505 CAS Number: 206055-79-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...