2'-((5-(4-Methylpiperazin-1-yl)pyridin-2-yl)amino)-7',8'-dihydro-6'H-spiro[cyclohexane-1,9'-pyrazino[1',2':1,5]pyrrolo[2,3-d]pyrimidin]-6'-one ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Trilaciclib
A982876-1MG
A982876-5MG
A982876-10MG
77599-334EA
124.03
USD
77599-334
77599-336
77623-252
2'-((5-(4-Methylpiperazin-1-yl)pyridin-2-yl)amino)-7',8'-dihydro-6'H-spiro[cyclohexane-1,9'-pyrazino[1',2':1,5]pyrrolo[2,3-d]pyrimidin]-6'-one ≥98%
2'-((5-(4-Methylpiperazin-1-yl)pyridin-2-yl)amino)-7',8'-dihydro-6'H-spiro[cyclohexane-1,9'-pyrazino[1',2':1,5]pyrrolo[2,3-d]pyrimidin]-6'-one
Precaución: for Research Use Only
Formula:
C₂₄H₃₀N₈O MW: 446.55 g/mol |
MDL Number:
MFCD30747890 CAS Number: 1374743-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...