1-(2-(2-(Mesitylimino)-9,10-dimethoxy-4-oxo-6,7-dihydro-2H-pyrimido[6,1-a]isoquinolin-3(4H)-yl)ethyl)urea ≥98%

Supplier: Ambeed
Warning

Synonyms: (E/Z)-Ensifentrine

A982733-1MG A982733-5MG
77599-332EA 21.53 USD
77599-332 77623-250
1-(2-(2-(Mesitylimino)-9,10-dimethoxy-4-oxo-6,7-dihydro-2H-pyrimido[6,1-a]isoquinolin-3(4H)-yl)ethyl)urea ≥98%
1-(2-(2-(Mesitylimino)-9,10-dimethoxy-4-oxo-6,7-dihydro-2H-pyrimido[6,1-a]isoquinolin-3(4H)-yl)ethyl)urea

Precaución: for Research Use Only

Formula: C₂₆H₃₁N₅O₄
MW: 477.56 g/mol
CAS Number: 298680-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR