Fmoc-D-allylglycine ≥95%

Supplier: Ambeed
Warning

Synonyms: D-2-Allylglycine, N-FMOC protected, Fmoc-D-Gly(allyl)-OH

A982634-1G A982634-250MG A982634-5G
77603-386EA 39.98 USD
77603-386 77603-388 77674-438
Fmoc-D-allylglycine ≥95%
Fmoc-D-allylglycine

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
MDL Number: MFCD01311776
CAS Number: 170642-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR