(5-Ethylthiophen-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A981710-1G A981710-100MG
77627-788EA 304.88 USD
77627-788 77878-771
(5-Ethylthiophen-2-yl)boronic acid ≥95%
(5-Ethylthiophen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₉BO₂S
MDL Number: MFCD02029241
CAS Number: 162607-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR