Rhenium trioxide 99.9% Re :79.5 ± 1.2%

Supplier: Ambeed
Warning

A981159-250MG A981159-100MG A981159-1G A981159-5G
77603-384EA 60.48 USD
77603-384 77670-430 77670-432 77670-434
Rhenium trioxide 99.9% Re :79.5 ± 1.2%
Rhenium trioxide

Precaución: for Research Use Only

Formula: O₃Re
MDL Number: MFCD00016306
CAS Number: 1314-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR