tert-Butyl 3-[(tetramethyl-1,3,2-dioxaborolan-2-yl)methylidene]azetidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A980554-1G A980554-250MG A980554-5G
77749-817EA 437 USD
77749-817 77749-818 77749-819
tert-Butyl 3-[(tetramethyl-1,3,2-dioxaborolan-2-yl)methylidene]azetidine-1-carboxylate ≥98%
tert-Butyl 3-[(tetramethyl-1,3,2-dioxaborolan-2-yl)methylidene]azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₆BNO₄
MW: 295.18 g/mol
MDL Number: MFCD31380980
CAS Number: 2246802-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR