(R)-5-Amino-1-methylpiperidin-2-one ≥98%

Supplier: Ambeed
Danger

A980415-100MG A980415-1G
77784-479EA 227 USD
77784-479 77784-480
(R)-5-Amino-1-methylpiperidin-2-one ≥98%
(R)-5-Amino-1-methylpiperidin-2-one

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MDL Number: MFCD30364322
CAS Number: 2002478-53-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR