1-(Phenylcarbamoyl)cyclopropanecarboxylic acid ≥95%

Supplier: Ambeed
Warning

A980010-100MG A980010-1G
77618-690EA 130.81 USD
77618-690 77618-692
1-(Phenylcarbamoyl)cyclopropanecarboxylic acid ≥95%
1-(Phenylcarbamoyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.2099 g/mol
MDL Number: MFCD16669493
CAS Number: 145591-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR