(R)-(4-(1-((tert-Butoxycarbonyl)amino)ethyl)phenyl)boronic acid ≥96%

Supplier: Ambeed
Warning

A979528-250MG A979528-5G A979528-100MG A979528-1G
77749-815EA 139 USD
77749-815 77749-816 77749-813 77749-814
(R)-(4-(1-((tert-Butoxycarbonyl)amino)ethyl)phenyl)boronic acid ≥96%
(R)-(4-(1-((tert-Butoxycarbonyl)amino)ethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₄
MDL Number: MFCD30834340
CAS Number: 871720-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR